accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 1731-94-8

IUPAC Name: methyl nonadecanoate
Synonyms: methyl nonadecanoate
Molecular Weight (g/mol): 312.54
Molecular Formula: C20H40O2
InChi Key: BDXAHSJUDUZLDU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC(=O)OC
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)