Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonadecanoic Acid Methyl Ester, ≥99%, Ultra Scientific

Catalog No. USFLSA055
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-FLSA-055 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-FLSA-055 Supplier Agilent Technologies Supplier No. FLSA055
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1731-94-8
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.538
InChI Key BDXAHSJUDUZLDU-UHFFFAOYSA-N
Synonym methyl nonadecan-1-oate, nonadecanoic acid methyl ester, nonadecanoic acid, methyl ester, unii-67fq8vv2l3, methylnonadecanoate, nonadecanoic acid methyl, nonadecanoic acid methylester, nonadecanoic acid,methyl ester, n-nonadecanoic acid methyl ester, 1n-nonadecanoic acid methyl ester
PubChem CID 15610
IUPAC Name methyl nonadecanoate
SMILES CCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

CAS Max % 99.93
Quantity 100 mg
Physical Form Liquid
Chemical Name or Material Nonadecanoic Acid Methyl Ester, 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.