Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Nonadecanoate 98.0+%, TCI America™
SDP

Catalog No. N046025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N046025G 25 g
N04605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N046025G Supplier TCI America Supplier No. N046025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1731-94-8
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.538
MDL Number MFCD00009011
InChI Key BDXAHSJUDUZLDU-UHFFFAOYSA-N
Synonym methyl nonadecan-1-oate, nonadecanoic acid methyl ester, nonadecanoic acid, methyl ester, unii-67fq8vv2l3, methylnonadecanoate, nonadecanoic acid methyl, nonadecanoic acid methylester, nonadecanoic acid,methyl ester, n-nonadecanoic acid methyl ester, 1n-nonadecanoic acid methyl ester
PubChem CID 15610
IUPAC Name methyl nonadecanoate
SMILES CCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 40°C
Color White
Quantity 25 g
Formula Weight 312.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Nonadecanoate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.