accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1735-17-7

IUPAC Name: (²H₁₂)cyclohexane
Synonyms: (²H₁₂)cyclohexane
Molecular Weight (g/mol): 96.24
Molecular Formula: C6H12
InChi Key: XDTMQSROBMDMFD-LBTWDOQPSA-N
SMILES: [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (1)