Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane-d12, for NMR, 99.5 atom % D, Thermo Scientific™

Catalog No. AC166280251
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC166280251 25 mL
AC166280051 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC166280251 Supplier Thermo Scientific Chemicals Supplier No. 166280251
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 1735-17-7
Molecular Formula C6H12
Molecular Weight (g/mol) 96.24
MDL Number 00044212
InChI Key XDTMQSROBMDMFD-LBTWDOQPSA-N
IUPAC Name (²H₁₂)cyclohexane
SMILES [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Quantity 25 mL
Water ≤0.03 %
Flash Point -18°C
Linear Formula C6D12
Chemical Name or Material Cyclohexane-d12
Hazard Category Exclamation mark
Hazard Statement Health hazard
EINECSNumber 217-077-3
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.