Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexane-d{12}, 99.5%(Isotopic)

Catalog No. AA42271SA
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42271SA 2 Ea.
AA42271KV 10 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42271SA Supplier Thermo Scientific Chemicals Supplier No. 042271SA
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1735-17-7 | C6H12 | 96.24 g/mol

It is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1735-17-7
Molecular Formula C6H12
Molecular Weight (g/mol) 96.24
MDL Number MFCD00044212
InChI Key XDTMQSROBMDMFD-LBTWDOQPSA-N
Synonym cyclohexane-d12, 2h12 cyclohexane, cyclohexane d, cyclohexane, d12, 2 h?? cyclohexane, cyclohexane-d12, ≥99.6 atom % d, cyclohexane #, cyclohexane-d12, 99.5 atom % d
PubChem CID 123129
SMILES [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Quantity 2 Ea.
Density 0.89 g/mL
Refractive Index 1.421
Flash Point -18°C (-1°F)
Melting Point 6.5°C
Boiling Point 81°C
UN Number UN1145
Beilstein 1908842
Solubility Information Insoluble in water.
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6-dodecadeuteriocyclohexane
Formula Weight 96.24
Percent Purity 99.5%
Chemical Name or Material Cyclohexane-d{12}, Isotopic
Show More Show Less
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEXANE
EINECSNumber 217-077-3
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.