CAS RN 17360-35-9
IUPAC Name:
tris((2E)-but-2-enedioic acid); bis(1-[4-(pyrrolidin-1-yl)but-2-yn-1-yl]pyrrolidin-2-one)
Synonyms:
tris(fumaric acid); bis(oxotremorine)
Molecular Weight (g/mol):
760.79
Molecular Formula:
C36H48N4O14
InChi Key:
WLYYOFJEBGHKEC-VQYXCCSOSA-N
SMILES:
OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.O=C1CCCN1CC#CCN1CCCC1.O=C1CCCN1CC#CCN1CCCC1
1
–
1
of
1
results
1
R&D Systems™ Oxotremorine sesquifumarate
CAS: 17360-35-9 Molecular Formula: C36H48N4O14