accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(Z)

CAS RN 17369-59-4

IUPAC Name: (3Z)-3-propylidene-1,3-dihydro-2-benzofuran-1-one
Synonyms: propylidenephthalide propylidene phthalide
Molecular Weight (g/mol): 174.2
Molecular Formula: C11H10O2
InChi Key: NGSZDVVHIGAMOJ-YHYXMXQVSA-N
SMILES: CC\C=C1/OC(=O)C2=CC=CC=C12
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)