accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBrBrNHNNBr

CAS RN 17374-26-4

IUPAC Name: 4,5,6,7-tetrabromo-2H-1,2,3-benzotriazole
Synonyms: 4,5,6,7-tetrabromo-2H-1,2,3-benzotriazole
Molecular Weight (g/mol): 434.71
Molecular Formula: C6HBr4N3
InChi Key: OMZYUVOATZSGJY-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(Br)=C(Br)C2=NNN=C12
Form 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results