accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3HNOCH3

CAS RN 17375-63-2

IUPAC Name: [2-(5-methoxy-2-phenyl-1H-indol-3-yl)ethyl]dimethylamine
Synonyms: [2-(5-methoxy-2-phenyl-1H-indol-3-yl)ethyl]dimethylamine
Molecular Weight (g/mol): 294.4
Molecular Formula: C19H22N2O
InChi Key: VSGPGYWZVPDDSK-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=C(CCN(C)C)C2=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)