accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1738-25-6

IUPAC Name: 3-(dimethylamino)propanenitrile
Synonyms: 3-(dimethylamino)propanenitrile
Molecular Weight (g/mol): 98.15
Molecular Formula: C5H10N2
InChi Key: MTPJEFOSTIKRSS-UHFFFAOYSA-N
SMILES: CN(C)CCC#N
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)