Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Dimethylaminopropionitrile 98.0+%, TCI America™
SDP

Catalog No. D0662500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0662500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0662500ML Supplier TCI America Supplier No. D0662500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1738-25-6
Molecular Formula C5H10N2
Molecular Weight (g/mol) 98.15
MDL Number MFCD00001957
InChI Key MTPJEFOSTIKRSS-UHFFFAOYSA-N
Synonym 3-dimethylamino propanenitrile, dimethylaminopropionitrile, 3-dimethylamino propionitrile, 3-dimethylaminopropionitrile, dmapn, beta-dimethylaminopropionitrile, propanenitrile, 3-dimethylamino, propionitrile, 3-dimethylamino, 3-n,n-dimethylamino propionitrile, n,n-dimethylamino-3-propionitrile
PubChem CID 15615
IUPAC Name 3-(dimethylamino)propanenitrile
SMILES CN(C)CCC#N

Specifications

Boiling Point 171°C
Quantity 500 mL
Formula Weight 98.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Dimethylaminopropionitrile
EINECSNumber (2)-1528
RTECSNumber UG1575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.