Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)propionitrile, 98%, Electrophoresis Grade, Thermo Scientific™

Catalog No. AAJ66058AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66058AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66058AK Supplier Thermo Scientific Chemicals Supplier No. J66058AK

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1738-25-6
Molecular Formula C5H10N2
Molecular Weight (g/mol) 98.15
MDL Number MFCD00001957
InChI Key MTPJEFOSTIKRSS-UHFFFAOYSA-N
Synonym 3-dimethylamino propanenitrile, dimethylaminopropionitrile, 3-dimethylamino propionitrile, 3-dimethylaminopropionitrile, dmapn, beta-dimethylaminopropionitrile, propanenitrile, 3-dimethylamino, propionitrile, 3-dimethylamino, 3-n,n-dimethylamino propionitrile, n,n-dimethylamino-3-propionitrile
PubChem CID 15615
IUPAC Name 3-(dimethylamino)propanenitrile
SMILES CN(C)CCC#N

Specifications

Melting Point -44°C
Color Colorless to Yellow
Density 0.870g/mL
Boiling Point 60°C to 61°C
Linear Formula (CH3)2NCH2CH2CN
Quantity 250 mL
Beilstein 773779
Formula Weight 98.15
Percent Purity 98%
Grade Electrophoresis
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)propionitrile
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H312-H315-H319-H335-H227
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber UG1575000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.