accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOONHOO

CAS RN 1738-86-9

IUPAC Name: 4-nitrophenyl 2-{[(benzyloxy)carbonyl]amino}acetate
Synonyms: 4-nitrophenyl 2-{[(benzyloxy)carbonyl]amino}acetate
Molecular Weight (g/mol): 330.3
Molecular Formula: C16H14N2O6
InChi Key: LHFNPUGRSYOPLF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(=O)CNC(=O)OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results