accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH

CAS RN 174063-99-1

IUPAC Name: pentadeca-2,4-diynoic acid
Synonyms: pentadeca-2,4-diynoic acid
Molecular Weight (g/mol): 234.34
Molecular Formula: C15H22O2
InChi Key: XRCJABKXJJKZPH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC#CC#CC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results