accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17413-10-4

IUPAC Name: (2,3-dihydro-1,4-benzodioxin-6-yl)methanaminium
Synonyms: 2,3-dihydro-1,4-benzodioxin-6-ylmethanaminium
Molecular Weight (g/mol): 166.2
Molecular Formula: C9H12NO2
InChi Key: FUDYRLUSXBRPIA-UHFFFAOYSA-O
SMILES: [NH3+]CC1=CC=C2OCCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results