accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHO

CAS RN 17419-81-7

IUPAC Name: 4-hydroxycyclohexane-1-carboxylic acid
Synonyms: 4-hydroxycyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: HCFRWBBJISAZNK-UHFFFAOYSA-N
SMILES: OC1CCC(CC1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results