accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CFSbFFFFF

CAS RN 174645-81-9

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium; hexafluorostibanuide
Synonyms: 1-butyl-3-methylimidazolium; hexafluorostibanuide
Molecular Weight (g/mol): 374.97
Molecular Formula: C8H15F6N2Sb
InChi Key: CSHAOLBENQQIBR-UHFFFAOYSA-H
SMILES: F[Sb-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results