accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNSOFFF

CAS RN 175202-93-4

IUPAC Name: 5-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carbaldehyde
Synonyms: 5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carbaldehyde
Molecular Weight (g/mol): 260.23
Molecular Formula: C10H7F3N2OS
InChi Key: ITUYPRXQRPMRTL-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C(F)(F)F)C1=CC=C(S1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results