CAS RN 175203-69-7
CAS RN 175203-69-7
IUPAC Name:
5-(2-phenylethynyl)pyridine-3-carboxylic acid
Synonyms:
5-(2-phenylethynyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol):
223.23
Molecular Formula:
C14H9NO2
InChi Key:
DXJZBANECHYHRT-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CN=CC(=C1)C#CC1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals 5-(2-Phenyleth-1-ynyl)nicotinic acid, 97%, Thermo Scientific™
CAS: 175203-69-7 Molecular Formula: C14H9NO2 Molecular Weight (g/mol): 223.231 MDL Number: MFCD00174055 InChI Key: DXJZBANECHYHRT-UHFFFAOYSA-N Synonym: 5-2-phenyleth-1-ynyl nicotinic acid,5-2-phenylethynyl pyridine-3-carboxylic acid,5-phenylethynyl pyridine-3-carboxylic acid,3-pyridinecarboxylicacid, 5-2-phenylethynyl,maybridge1_004925,5-phenylethynylnicotinic acid,5-phenylethynyl-nicotinic acid,5-phenylethynyl nicotinic acid,5-phenylethynyl-pyridine-3-carboxylic acid PubChem CID: 2779236 IUPAC Name: 5-(2-phenylethynyl)pyridine-3-carboxylic acid SMILES: C1=CC=C(C=C1)C#CC2=CC(=CN=C2)C(=O)O