accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 175277-45-9

H3CONFFF

CAS RN 175277-45-9

IUPAC Name: 2-methoxy-5-(trifluoromethyl)pyridine
Synonyms: 2-methoxy-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 177.13
Molecular Formula: C7H6F3NO
InChi Key: KNIGTEGBOBDGKP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=N1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results