Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. M283225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M283225G 25 g
M28325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M283225G Supplier TCI America Supplier No. M283225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175277-45-9
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00153203
InChI Key KNIGTEGBOBDGKP-UHFFFAOYSA-N
PubChem CID 2775312
IUPAC Name 2-methoxy-5-(trifluoromethyl)pyridine
SMILES COC1=NC=C(C=C1)C(F)(F)F

Specifications

Color Colorless
Quantity 25 g
UN Number 1993
Formula Weight 177.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.