accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOFFF

CAS RN 175278-07-6

IUPAC Name: [2-(trifluoromethoxy)phenyl]methanol
Synonyms: [2-(trifluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 192.14
Molecular Formula: C8H7F3O2
InChi Key: ICOVMLDFMWLRJO-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1OC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results