accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17573-92-1

IUPAC Name: 3-methoxythiophene
Synonyms: p-methoxythiophene
Molecular Weight (g/mol): 114.16
Molecular Formula: C5H6OS
InChi Key: RFSKGCVUDQRZSD-UHFFFAOYSA-N
SMILES: COC1=CSC=C1
Boiling Point 
  • (2)
  • (1)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (4)
  • (2)
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
  • (3)
  • (5)
  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (3)
  • (6)