Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxythiophene, 98%

Catalog No. AC381680250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC381680250 25 g Glass bottle
AC381680010 1 g Glass bottle
AC381680050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC381680250 Supplier Thermo Scientific Chemicals Supplier No. 381680250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 17573-92-1 | C5H6OS | 114.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17573-92-1
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.17
MDL Number MFCD00043894
InChI Key RFSKGCVUDQRZSD-UHFFFAOYSA-N
Synonym thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411
PubChem CID 140249
IUPAC Name 3-methoxythiophene
SMILES COC1=CSC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Density 1.1430g/mL
Boiling Point 80.0°C to 82.0°C (65.0 mmHg)
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5300 to 1.5320 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.143
Formula Weight 114.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxythiophene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.