accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17586-89-9

NSSNOONOO

CAS RN 17586-89-9

IUPAC Name: 2-[(2,4-dinitrophenyl)sulfanyl]-1,3-benzothiazole
Synonyms: 2-[(2,4-dinitrophenyl)sulfanyl]-1,3-benzothiazole
Molecular Weight (g/mol): 333.34
Molecular Formula: C13H7N3O4S2
InChi Key: RCPUUVXIUIWMEE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(SC2=NC3=CC=CC=C3S2)C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)