CAS RN 1762-95-4
Ammonium Thiocyanate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 MDL Number: MFCD00011428 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium thiocyanate, 99+%, extra pure
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.12 MDL Number: MFCD00011428 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium thiocyanate, 98%, ACS reagent
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.12 MDL Number: MFCD00011428 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium thiocyanate, 98+%
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 MDL Number: MFCD00011428 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium thiocyanate, ACS, 97.5% min
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 MDL Number: MFCD00011428 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium Thiocyanate, Crystal, Reagent, ACS, 97.5%, Spectrum™ Chemical
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.12 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N IUPAC Name: ammonium cyanosulfanide SMILES: [NH4+].[S-]C#N
Ammonium thiocyanate, 99.99% trace metals basis, MilliporeSigma™ Supelco™
CAS: 1762-95-4 Molecular Weight (g/mol): 76.12 MDL Number: MFCD00011428 Synonym: Ammonium rhodanide
Ammonium Thiocyanate, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium Thiocyanate, MilliporeSigma™
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium Thiocyanate TS, (U.S.P. Test Solution), Spectrum™ Chemical
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.12 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N IUPAC Name: ammonium cyanosulfanide SMILES: [NH4+].[S-]C#N
Ammonium Thiocyanate, ACS Reagent Grade, Ricca Chemical
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium Thiocyanate TS, 8% (w/v), Ricca Chemical
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium Thiocyanate 98.0+%, TCI America™
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 MDL Number: MFCD00011428 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
Ammonium Thiocyanate, 0.0250 N (N/40), Ricca Chemical
CAS: 1762-95-4 Molecular Formula: CH4N2S Molecular Weight (g/mol): 76.117 InChI Key: SOIFLUNRINLCBN-UHFFFAOYSA-N Synonym: ammonium thiocyanate,ammoniumrhodanid,thiocyanic acid, ammonium salt,ammonium isothiocyanate,ammonium rhodanate,weedazol tl,trans-aid,ammonium sulfocyanate,ammonium sulfocyanide,rhodanine, ammonium salt PubChem CID: 15666 IUPAC Name: azanium;thiocyanate SMILES: C(#N)[S-].[NH4+]
- 1
- 2