Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Thiocyanate, MilliporeSigma™

Catalog No. M1012130500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1012130500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1012130500 Supplier MilliporeSigma Supplier No. 1.01213.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Physical Form Solid
Quantity 500 g
Solubility Information Soluble in water: 1600g/L at 20°C
Formula Weight 76.12
Percent Purity ≥99.0%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Vapor Pressure <1hPa at 20°C
Density 1.3g/cm3 at 20°C
Chemical Name or Material Ammonium Thiocyanate
Show More Show Less
Hazard Category H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
Hazard Statement H412: Harmful to aquatic life with long lasting effects.
EINECSNumber 217-175-6
RTECSNumber XK7875000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.