Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium thiocyanate, 99.99% trace metals basis, MilliporeSigma™ Supelco™

Catalog No. 111018914 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8914 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8914 Supplier MilliporeSigma Supelco Supplier No. 43135450G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium thiocyanate is an ammonium salt commonly used for thiocyanation reaction of organic compounds.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Weight (g/mol) 76.12
MDL Number MFCD00011428
Synonym Ammonium rhodanide

Specifications

Melting Point 152°C to 154°C
Physical Form Crystals
Quantity 50 g
Packaging Poly Bottle
Linear Formula NH4 SCN
Solubility Information Water: 76.1 g/L (at 20°C)
Formula Weight 76.12
Percent Purity 99.99% (trace metals basis)
Grade ACS
Chemical Name or Material Ammonium Thiocyanate
RTECSNumber XK7875000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.