CAS RN 17650-98-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Caerulein, Sulfated, MP Biomedicals™
Supplied as a trifluoroacetate salt. Caerulein, Sulfated, MP Biomedicals is an antagonist of endogenous opiods and a gastroregulatory molecule with actions similar to cholecystokinin. It prevents gall bladder pain, renal colic, and the pain of intermitten
2
Tocris Bioscience™ Caerulein
CCK agonist