accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOFN

CAS RN 176508-81-9

IUPAC Name: 4-cyano-3-fluorobenzoic acid
Synonyms: 4-cyano-3-fluorobenzoic acid
Molecular Weight (g/mol): 165.12
Molecular Formula: C8H4FNO2
InChi Key: ZWKNDLMYSLLMRF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results