accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1766-76-3

IUPAC Name: bis(2,3,4,5,6-pentafluorophenyl)methanol
Synonyms: bis(2,3,4,5,6-pentafluorophenyl)methanol
Molecular Weight (g/mol): 364.14
Molecular Formula: C13H2F10O
InChi Key: WRLLBTKDSCJOBL-UHFFFAOYSA-N
SMILES: OC(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)