Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decafluorobenzhydrol 99.0+%, TCI America™
SDP

Catalog No. D16415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D16415G Supplier TCI America Supplier No. D16415G

Chemical Identifiers

CAS 1766-76-3
Molecular Formula C13H2F10O
Molecular Weight (g/mol) 364.14
MDL Number MFCD00000297
InChI Key WRLLBTKDSCJOBL-UHFFFAOYSA-N
Synonym decafluorobenzhydrol, bis pentafluorophenyl methanol, bis perfluorophenyl methanol, bis 2,3,4,5,6-pentafluorophenyl methanol, 2,3,4,5,6,2',3',4',5',6'-decafluorobenzhydrol, benzhydrol, 2,2',3,3',4,4',5,5',6,6'-decafluoro, benzenemethanol, 2,3,4,5,6-pentafluoro-.alpha.-pentafluorophenyl, bis 2,3,4,5,6-pentafluorophenyl methanol #, bis 2,3,4,5,6-pentafluorophenyl methan-1-ol, 2,3,4,5,6,2',3',4',5',6'-decafluorobenzylhydrol
PubChem CID 74485
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)methanol
SMILES OC(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 80°C
Color White-Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 364.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Decafluorobenzhydrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.