Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decafluorobenzhydrol, 97%

Catalog No. AAL0219706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0219706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0219706 Supplier Thermo Scientific Chemicals Supplier No. L0219706

missing translation for 'validMainframeMsg'

CAS: 1766-76-3 | C13H2F10O | 364.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1766-76-3
Molecular Formula C13H2F10O
Molecular Weight (g/mol) 364.14
MDL Number MFCD00000297
InChI Key WRLLBTKDSCJOBL-UHFFFAOYSA-N
Synonym decafluorobenzhydrol, bis pentafluorophenyl methanol, bis perfluorophenyl methanol, bis 2,3,4,5,6-pentafluorophenyl methanol, 2,3,4,5,6,2',3',4',5',6'-decafluorobenzhydrol, benzhydrol, 2,2',3,3',4,4',5,5',6,6'-decafluoro, benzenemethanol, 2,3,4,5,6-pentafluoro-.alpha.-pentafluorophenyl, bis 2,3,4,5,6-pentafluorophenyl methanol #, bis 2,3,4,5,6-pentafluorophenyl methan-1-ol, 2,3,4,5,6,2',3',4',5',6'-decafluorobenzylhydrol
PubChem CID 74485
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)methanol
SMILES OC(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Beilstein 2067248
Solubility Information Insoluble in water.
Formula Weight 364.14
Percent Purity 97%
Chemical Name or Material Decafluorobenzhydrol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 217-185-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.