accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3CCH3NCS

CAS RN 17701-76-7

IUPAC Name: 2-isothiocyanato-2,4,4-trimethylpentane
Synonyms: 2-isothiocyanato-2,4,4-trimethylpentane
Molecular Weight (g/mol): 171.3
Molecular Formula: C9H17NS
InChi Key: KUGHMFIBHMZICO-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)N=C=S
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results