CAS RN 177172-49-5
IUPAC Name:
hydrogen [3-(2-chlorophenyl)propyl][(1R)-1-(3-methoxyphenyl)ethyl]amine chloride
Synonyms:
hydrogen [3-(2-chlorophenyl)propyl][(1R)-1-(3-methoxyphenyl)ethyl]amine chloride
Molecular Weight (g/mol):
340.29
Molecular Formula:
C18H23Cl2NO
InChi Key:
YJXUXANREVNZLH-PFEQFJNWSA-N
SMILES:
[H+].[Cl-].COC1=CC=CC(=C1)[C@@H](C)NCCCC1=CC=CC=C1Cl
1
–
1
of
1
results
1
R&D Systems™ R 568 hydrochloride
CAS: 177172-49-5 Molecular Formula: C18H23Cl2NO