accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNH2(R)OHO(R)OH(R)HO(R)HOH

CAS RN 1772-03-8

IUPAC Name: hydrogen (3R,4R,5R,6R)-3-amino-6-(hydroxymethyl)oxane-2,4,5-triol chloride
Synonyms: hydrogen galactosamine chloride
Molecular Weight (g/mol): 215.63
Molecular Formula: C6H14ClNO5
InChi Key: QKPLRMLTKYXDST-BMZZJELJSA-N
SMILES: [H+].[Cl-].N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
Physical Form 
  • (3)
Quantity 
  • (2)
  • (1)
Form 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)