Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Galactosamine hydrochloride, 99%, MP Biomedicals™

Catalog No. ICN10174305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10174305 5 g
ICN10174301 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10174305 Supplier MP Biomedicals, Inc Supplier No. 0210174305
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1772-03-8
Molecular Formula C6H13NO5·HCl
Synonym Chondrosamine hydrochloride, 2-Amino-2-deoxy-D-galactopyranose hydrochloride

Specifications

Color White
Assay Percent Range 0.99
Quantity 5 g
Formula Weight 215.64
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Galactosamine hydrochloride
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.