Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Galactosamine Hydrochloride, TCI America™
SDP

Catalog No. G00071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G00071G Supplier TCI America Supplier No. G00071G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1772-03-8
Molecular Formula C6H13NO5
MDL Number MFCD00135830
Synonym Chondrosamine Hydrochloride

Specifications

Melting Point 176°C
Color White
Quantity 1 g
Formula Weight 215.63
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Galactosamine Hydrochloride
RTECSNumber LW5500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.