accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNHF

CAS RN 177858-80-9

IUPAC Name: 7-fluoro-1H,2H,3H,4H,9H-pyrido[3,4-b]indole
Synonyms: 7-fluoro-1H,2H,3H,4H,9H-pyrido[3,4-b]indole
Molecular Weight (g/mol): 190.22
Molecular Formula: C11H11FN2
InChi Key: OTGGOPSBNVEBDA-UHFFFAOYSA-N
SMILES: FC1=CC=C2C3=C(CNCC3)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results