accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNFOHO

CAS RN 177966-64-2

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-fluorophenyl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoic acid
Molecular Weight (g/mol): 405.43
Molecular Formula: C24H20FNO4
InChi Key: IXUMACXMEZBPJG-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=C(F)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results