Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-4-fluoro-D-phenylalanine, 95%, Thermo Scientific™

Catalog No. AAH6692406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6692406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6692406 Supplier Thermo Scientific Chemicals Supplier No. H6692406

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177966-64-2
Molecular Formula C24H20FNO4
Molecular Weight (g/mol) 405.425
MDL Number MFCD00235900
InChI Key IXUMACXMEZBPJG-JOCHJYFZSA-N
Synonym fmoc-d-phe 4-f-oh, fmoc-d-4-fluorophenylalanine, fmoc-4-fluoro-d-phenylalanine, fmoc-d-4-fluorophe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-fluorophenyl propanoic acid, fmoc-4-fluoro-d-phe, r-n-fmoc-4-fluorophenylalanine, fmoc-p-fluoro-d-phe-oh, n-alpha-9-fluorenylmethyloxycarbonyl-4-fluoro-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-fluoro
PubChem CID 16213164
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-fluorophenyl)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)F)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 405.43
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-4-fluoro-D-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.