CAS RN 1780-27-4
IUPAC Name:
4,5,6-trichloropyrimidine
Synonyms:
4,5,6-trichloropyrimidine
Molecular Weight (g/mol):
183.42
Molecular Formula:
C4HCl3N2
InChi Key:
AUWPHGWEYHEAIG-UHFFFAOYSA-N
SMILES:
ClC1=NC=NC(Cl)=C1Cl
1
–
1
of
1
results
1
4,5,6-Trichloropyrimidine 98.0+%, TCI America™
CAS: 1780-27-4 Molecular Formula: C4HCl3N2 Molecular Weight (g/mol): 183.42 MDL Number: MFCD08458457 InChI Key: AUWPHGWEYHEAIG-UHFFFAOYSA-N PubChem CID: 249007 IUPAC Name: 4,5,6-trichloropyrimidine SMILES: ClC1=NC=NC(Cl)=C1Cl