Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5,6-Trichloropyrimidine 98.0+%, TCI America™
SDP

Catalog No. T25501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25501G Supplier TCI America Supplier No. T25501G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1780-27-4
Molecular Formula C4HCl3N2
Molecular Weight (g/mol) 183.42
MDL Number MFCD08458457
InChI Key AUWPHGWEYHEAIG-UHFFFAOYSA-N
PubChem CID 249007
IUPAC Name 4,5,6-trichloropyrimidine
SMILES ClC1=NC=NC(Cl)=C1Cl

Specifications

Melting Point 53°C
Boiling Point 87°C
Quantity 1 g
Formula Weight 183.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5,6-Trichloropyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.