CAS RN 1781882-65-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Pyridostatin pentahydrochloride
Stabilizes G-quadruplexes; induces DNA damage and cell cycle arrest