CAS RN 1781882-72-1
IUPAC Name:
N-{[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethyl-1H-imidazo[4,5-c]pyridin-7-yl]methyl}piperidin-4-amine dihydrochloride
Synonyms:
N-{[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethylimidazo[4,5-c]pyridin-7-yl]methyl}piperidin-4-amine dihydrochloride
Molecular Weight (g/mol):
415.32
Molecular Formula:
C16H24Cl2N8O
InChi Key:
NRRCQHARLPNLHJ-UHFFFAOYSA-N
SMILES:
Cl.Cl.CCN1C(=NC2=CN=CC(CNC3CCNCC3)=C12)C1=NON=C1N
1
–
1
of
1
results
1
R&D Systems™ SB 747651A dihydrochloride
CAS: 1781882-72-1 Molecular Formula: C16H24Cl2N8O