CAS RN 1785666-59-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
R&D Systems™ CP 465022 hydrochloride
CAS: 1785666-59-2 Molecular Formula: C26H25Cl2FN4O