accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiOCH3OOCH3OOCH3O

CAS RN 17865-07-5

IUPAC Name: bis(acetyloxy)(propyl)silyl acetate
Synonyms: bis(acetyloxy)(propyl)silyl acetate
Molecular Weight (g/mol): 248.31
Molecular Formula: C9H16O6Si
InChi Key: DKGZKEKMWBGTIB-UHFFFAOYSA-N
SMILES: CCC[Si](OC(C)=O)(OC(C)=O)OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results