accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH2

CAS RN 17878-39-6

IUPAC Name: ethenyl(methyl)phenylsilyl
Synonyms: ethenyl(methyl)phenylsilyl
Molecular Weight (g/mol): 147.27
Molecular Formula: C9H11Si
InChi Key: MOSXLDGILGBOSZ-UHFFFAOYSA-N
SMILES: C[Si](C=C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results