accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCHOOCH3

CAS RN 17920-23-9

IUPAC Name: 1,3-diethyl 2-(prop-2-yn-1-yl)propanedioate
Synonyms: 1,3-diethyl 2-(prop-2-yn-1-yl)propanedioate
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: FKXOELIKVUIBLP-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC#C)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results